Learn More
Research Products International Corp Staining Dish, 94 X 81 X 49 mm
SDP
Catalog No. 50212950
Encompass
Staining Dish has a light-tight cover with a compartment for drying agents. Dish also features a perforated slide partition that supports the drying agent. Slide partition has index numbers and writing surface. This small size dish saves on staining liquids, developing fluids and intermediates since only 150ml is needed. The dish is easy to clean and especially suited for storing enzymes.For use with Staining Rack.
Catalog No. 50-212-950 Supplier Research Products International Corp Supplier No. F44182
Staining Dish has a light-tight cover with a compartment for drying agents. Dish also features a perforated slide partition that supports the drying agent. Slide partition has index numbers and writing surface. This small size dish saves on staining liquids, developing fluids and intermediates since only 150ml is needed. The dish is easy to clean and especially suited for storing enzymes.For use with Staining Rack.