Learn More
Med Vet International WOW - World of Wonder Microscope
Catalog No. 503046148
Encompass_Preferred
View 3D objects up close! Lightweight, top and bottom LED lights for solid objects and prepared slides, glass optics, 10x and 25x eyepieces for up to 50x total magnification. Includes 5 specimens with slide carrying case and experiment guide.
Catalog No. 50-304-6148 Supplier Med Vet International Supplier No. SMD09
View 3D objects up close! Lightweight, top and bottom LED lights for solid objects and prepared slides, glass optics, 10x and 25x eyepieces for up to 50x total magnification. Includes 5 specimens with slide carrying case and experiment guide.