Learn More
LabScientific Poly-L-Lysine coated adhesive microscope slides. 1"X3", 1mm thick. RNase Free, Positively Charged. 72 slides/pack.
Catalog No. NC9202659
Encompass
Poly-L-Lysine Coated Adhesive Microscope Slides, RNase Free, Positively Charged, Clear with White Frosted Edge, Clean Room Prepared. Prevent loss of paraffin embedded and frozen sections for histologic staining 72 slides/pack.
Catalog No. NC9202659 Supplier LabScientific Supplier No. 7799
Poly-L-Lysine Coated Adhesive Microscope Slides, RNase Free, Positively Charged, Clear with White Frosted Edge, Clean Room Prepared. Prevent loss of paraffin embedded and frozen sections for histologic staining 72 slides/pack.