Learn More
Waters Corp Waters Corporation Microscopic Slide Holder, MALDI SYNAPT XS Compatible, for Imaging Experiments
Catalog No. 50792926 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This microscopic slide holder, also known as a glass slide adapter, is designed for use with the MALDI SYNAPT XS system. It serves as a crucial sample plate format, particularly for imaging experiments, and is one of the various predefined formats compatible with the system.
  • Designed for the MALDI SYNAPT XS system.
  • Compatible with all Waters sample plate formats.
  • Functions as a glass slide adapter.
  • Ideal for imaging experiments.
  • Selected using Waters HDI software for imaging applications.
Catalog No. 50-792-926 Supplier Waters Corp Supplier No. 700003896
This microscopic slide holder, also known as a glass slide adapter, is designed for use with the MALDI SYNAPT XS system. It serves as a crucial sample plate format, particularly for imaging experiments, and is one of the various predefined formats compatible with the system.
  • Designed for the MALDI SYNAPT XS system.
  • Compatible with all Waters sample plate formats.
  • Functions as a glass slide adapter.
  • Ideal for imaging experiments.
  • Selected using Waters HDI software for imaging applications.