Learn More
Bio Rad Laboratories MICROSEAL 384-WELL PCR PLATES
Catalog No. 509011612
Encompass
PCR plates, 384-well Bio-Rad Skirted Version 2.0 White PCR volumes of 5 to 25uL are recommended Polypropylene Raised rim Free of DNase, RNase, and human DNA Suitable for high-throughput thermal-cycling applications, 50
Catalog No. 50-901-1612 Supplier Bio Rad Laboratories Supplier No. MSP3852
PCR plates, 384-well Bio-Rad Skirted Version 2.0 White PCR volumes of 5 to 25uL are recommended Polypropylene Raised rim Free of DNase, RNase, and human DNA Suitable for high-throughput thermal-cycling applications, 50