Learn More
Bio Rad Laboratories MICROSEAL 384-WELL PCR PLATES
Supplier:  Bio Rad Laboratories MSP3862
Encompass
PCR plates, 384-well Bio-Rad Skirted Version 2.0 Black PCR volumes of 5 to 25uL are recommended Polypropylene Raised rim Free of DNase, RNase, and human DNA Suitable for high-throughput thermal-cycling applications, 50
Catalog No. 50-901-1613