Learn More
Bio Rad Laboratories MICROSEAL 96-WELL PCR PLATES
Catalog No. 509011811
Encompass
PCR plate, 96-well Bio-Rad Microseal Skirted Blue Polypropylene Recommended PCR volumes 5 to 125uL Raised rim Free of DNase, RNase, and human DNA Suitable for high-throughput thermal-cycling applications 50
Catalog No. 50-901-1811 Supplier Bio Rad Laboratories Supplier No. MSP9631
Add to Cart
Add to Cart
PCR plate, 96-well Bio-Rad Microseal Skirted Blue Polypropylene Recommended PCR volumes 5 to 125uL Raised rim Free of DNase, RNase, and human DNA Suitable for high-throughput thermal-cycling applications 50