Learn More
Bio Rad Laboratories MICROSEAL 96WELL PCR PLTE 50PK
Catalog No. 509011614
Encompass
PCR plates, 96-well Bio-Rad Skirted Natural PCR volumes of 5 to 125uL are recommended Polypropylene Raised rim Free of DNase, RNase, and human DNA Suitable for high-throughput thermal-cycling applications 50
Catalog No. 50-901-1614 Supplier Bio Rad Laboratories Supplier No. MSP9601
PCR plates, 96-well Bio-Rad Skirted Natural PCR volumes of 5 to 125uL are recommended Polypropylene Raised rim Free of DNase, RNase, and human DNA Suitable for high-throughput thermal-cycling applications 50