Learn More
Bio Rad Laboratories MICROSEAL A FILM 50/PK
Catalog No. 509010035
Encompass
Microseal A Film; Bio-Rad; Compliant inner layer designed to seal tightly during PCR and release smoothly; Microseal A film not designed for optical use; Do not use this film in real-time PCR detection systems; Package of 50 seals
Catalog No. 50-901-0035 Supplier Bio Rad Laboratories Supplier No. MSA5001
Microseal A Film; Bio-Rad; Compliant inner layer designed to seal tightly during PCR and release smoothly; Microseal A film not designed for optical use; Do not use this film in real-time PCR detection systems; Package of 50 seals