Learn More
Bio Rad Laboratories MICROSEAL F FOILMICROSEAL F
Catalog No. 509010077
Encompass
Foil; Bio-Rad; Microseal F aluminized foil; Holds PCR volumes > /=25uL for 96-well or > /=5uL for 384-well; Can be pierced with pipets; Free of RNase, DNase, human DNA; Package of 100 seals
Catalog No. 50-901-0077 Supplier Bio Rad Laboratories Supplier No. MSF1001B
Foil; Bio-Rad; Microseal F aluminized foil; Holds PCR volumes > /=25uL for 96-well or > /=5uL for 384-well; Can be pierced with pipets; Free of RNase, DNase, human DNA; Package of 100 seals