Learn More
Berkshire™ MicroSeal SuperSorb™ Lite
Catalog No. 19070023 Shop All Berkshire Corporation Products
Change view
Click to view available options
Material:
Polyester Knit (1%)
Catalog No. Material
19-070-023 Polyester Knit (1%)
1 options
Catalog No. 19-070-023 Supplier Berkshire™ Supplier No. SSLT1018B10
Add to Cart
Add to Cart
MicroSeal Lite is the highest absorbency polyester knit wiper available on the market. Its unique design provides texture and extra durability when cleaning stubborn residues. It is an excellent wiper for applications where high concentrations of liquid are in use.
MicroSeal Lite is the highest absorbency polyester knit wiper available on the market. Its unique design provides texture and extra durability when cleaning stubborn residues. It is an excellent wiper for applications where high concentrations of liquid are in use.
  • Critically low particles, fibers, ions and extractables
  • Light weight, high absorbency material design reduces actual cost in use compared to heavier weight products
  • High abrasion resistance
  • Chemically compatible with IPA, acetone and other solvents
  • Pinsonic design adds surface texture that aids in stubborn residue removal
Specifications
Product Type Cleanroom Flat Mop Wiper
Sterility Non-sterile
Autoclavable Autoclavable
Color-coded No
Material Polyester Knit (1%)