Learn More
Cytiva (Formerly Pall Lab) Microsep Advance Centrifugal Devices with Omega Membrane - 1K, yellow (100/pkg)
Supplier:  Cytiva MCP001C46
Encompass
Precise, Quick Recovery of Microliter Volumes-Rapidly concentrates 0.5-5 mL sample volumes to 65100 L.-Achieve 50X concentration and > 90% recovery in just minutes-Deadstop feature prevents samples from spinning to dryness -Versatile Omega™ membrane is available in a variety of MWCOs-Color-coded and laser etched for easy identification This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9552