Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroSEQ™ 500 16S rDNA Sequencing Kit
GREENER_CHOICE

Catalog No. 4346480
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-464-80 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-464-80 Supplier Applied Biosystems™ Supplier No. 4346480
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The sequencing component of the MicroSEQ™ 500 16S rDNA Bacterial Identification System, which provides an easy-to-use DNA sequence-based method to identify most bacteria.

  • The sequencing component of the MicroSEQ™ 500 16S rDNA Bacterial Identification System, which provides an easy-to-use DNA sequence-based method to identify most bacteria
  • Use this kit to sequence PCR products that have been generated using the MicroSEQ™ 500 16S rDNA PCR Kit
  • The two kits together permit the quick and convenient amplification and sequencing of 500 base pairs of the 16S rRNA gene
  • Sequences can then be compared to the MicroSEQ™ 16S rDNA 500 Library to generate an identity match
  • This kit does not include a protocol or a Quick Reference Card
  • For Research Use Only. Not for use in diagnostics procedures.
  • Store at -15° to -25°C

Benefits of the MicroSEQ™ 500 16S rDNA Sequencing Kit

  • Sequence-based measurement ensures you get reproducible results in under 24 hours
  • Growth requirement-independent measurement enables you to identify bacteria using objective criteria
  • Universal domain-level primers eliminate up-front optimization
  • MicroSEQ™ ID Analysis Software ensures accurate species identification
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms
  • Requires minimal training-same work flow as the MicroSEQ™ D2 LSU fungal kit

Specifications

Content And Storage Each kit contains:
Two tubes each of forward and reverse sequencing mix. Sufficient reagent to perform 55 forward and 55 reverse sequencing reactions. Store at -15 to -25°C.
Detection Method Capillary Electrophoresis
Format Kit
Sample Type PCR Products
For Use With (Equipment) 310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Sequencing
Includes Forward sequencing mix (2 tubes), and reverse sequencing mix (2 tubes)
Libraries Customizable, Bacterial Library (> 2000 species)
No. of Reactions 55 Forward, 55 Reverse
Product Line MicroSEQ
Product Type Sequencing Kit
Quantity 1 Kit
Software MicroSEQ™ ID Analysis Software
Shipping Condition Dry Ice
Target 1st 500 bp of 16S rRNA gene
Target Organism Class Bacteria
Technique SBT (Sequence-Based Typing)
Workflow Step Cycle Sequencing
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.