Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroSEQ™ D2 rDNA Fungal Sequencing Kit
GREENER_CHOICE

Catalog No. 4347481
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-474-81 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-474-81 Supplier Applied Biosystems™ Supplier No. 4347481
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit is the sequencing component of the MicroSEQ Fungal Identification System, which provides an easy-to-use DNA sequence-based method to identify most fungi.

This kit is the sequencing component of the MicroSEQ™ Fungal Identification System, which provides an easy-to-use DNA sequence-based method to identify most fungi. It includes the primers needed to sequence the PCR products generated using the PCR component. A protocol and Quick Reference Card are not included.

  • Sequence-based measurement ensures you get reproducible results in less than 24 hours.
  • Growth requirement-independent measurement enables you to identify fungi using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training; same work flow as the MicroSEQ™ Full Gene 16S rDNA Bacterial Identification Kit.

Specific and Easy to Use
Based on phylogenetic analysis of the D2 region of large-subunit ribosomal RNA gene sequences, this easy-to-use kit enables you to identify even the most difficult fungi, yeast, and oomycete plant pathogens. To ensure accurate and specific results, the kit's software compares your samples to a database of D2 sequences from over 1000 validated type species.

The MicroSEQ™ ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems™ MicroSEQ™ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

Specifications

Content And Storage The kit contains reagents sufficient for 55 sequencing reactions:
• D2 Forward Sequencing Mix
• D2 Reverse Sequencing Mix

Store at -15°C to -25°C
Detection Method Capillary Electrophoresis
Format Kit
Sample Type PCR Products
For Use With (Equipment) 310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 9800 Fast Thermal Cycler
For Use With (Application) Sequencing
Libraries Fungal Library (> 1100 species), Customizable
Product Line MicroSEQ
Product Type Sequencing Kit
Quantity 1 Kit
Software MicroSEQ™ ID Analysis Software
Shipping Condition Dry Ice
Target D2 region of rRNA gene
Target Organism Class Fungi
Technique SBT (Sequence-Based Typing)
Workflow Step Cycle Sequencing
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.