Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
GREENER_CHOICE

Catalog No. 4347484
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-474-84 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-474-84 Supplier Applied Biosystems™ Supplier No. 4347484
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MicroSEQ Full Gene 16S rDNA Sequencing Kit is the sequencing component of the MicroSEQ 16S rDNA Full Gene Identification System, which provides an easy-to-use DNA sequence-based method to identify most bacteria.

The MicroSEQ™ Full Gene 16S rDNA Sequencing Kit is the sequencing component of the MicroSEQ™ 16S rDNA Full Gene Identification System, which provides an easy-to-use DNA sequence-based method to identify most bacteria. Use this kit to sequence PCR products that have been generated using the MicroSEQ™ Full Gene 16S rDNA PCR Kit. The two kits together permit the quick and convenient amplification and sequencing of the bacterial 16S rRNA gene. This kit includes a Protocol and a Quick Reference Card. The sequencing kit does not include a protocol or a Quick Reference Card.

  • Sequence-based measurement ensures you get reproducible results in under 24 hours.
  • Growth requirement-independent measurement enables you to identify bacteria using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training-similar work flow as the MicroSEQ™ D2 LSU fungal kit and the MicroSeq™ 500 16S rDNA kit.

The MicroSEQ™ ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems™ MicroSEQ™ System combines the advantages of MicroSEQ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

Specifications

Content And Storage Each kit contains three tubes each of forward and reverse sequencing mix. Sufficient reagent is supplied to perform 15 forward and 15 reverse sequencing reactions of the 16s rRNA gene.

Store at -15°C to -25°C.
Detection Method Capillary Electrophoresis
Format Kit
Sample Type PCR Products
For Use With (Equipment) 310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Sequencing
Libraries Customizable, Bacterial Library (> 2000 species)
Product Line MicroSEQ
Product Type Sequencing Kit
Quantity 1 Kit
Software MicroSEQ™ ID Analysis Software
Shipping Condition Dry Ice
Target 16S rRNA (full gene)
Target Organism Class Bacteria
Technique SBT (Sequence-Based Typing)
Workflow Step Cycle Sequencing
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.