Learn More
Applied Biosystems™ MicroSEQ™ ID Fungal Gene Library v2022
Catalog No. A57314
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
A57314 1 Ea.
1 options
Catalog No. A57314 Supplier Applied Biosystems™ Supplier No. A57314
Add to Cart
Add to Cart
Request a Quote
The MicroSEQ ID Fungal Gene Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0.
The MicroSEQ ID Fungal Gene Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0. It contains over 1300 reference sequences from the D2 region of the Large Subunit (rDNA) of yeast and mold species against which an unknown yeast or mold sequence can be compared in order to identify the unknown organism.
Specifications
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers
For Use With (Application) Microbial Identification
License Static
Operating System Microsoft Windows 10
Product Line MicroSeq™ ID
Product Type Microbial Identification Software
Quantity 1 Ea.
Software Data Collection and Analysis Software
What is the difference between MicroSEQ ID Fungal Gene Library v2018 and v2022?
MicroSEQ ID Fungal Gene Library v2022 contains identical content to the previous generation library, MicroSEQ ID Fungal Gene Library v2018. MicroSEQ ID Fungal Gene Library v2022 is compatible with MicroSEQ ID Microbial Identification Software v4.0 whereas MicroSEQ ID Fungal Gene Library v2018 is compatible with MicroSEQ ID Microbial Identification Software v3.1.3.
What is the browser compatibility requirement for MicroSEQ ID Fungal Gene Library v2022?
MicroSEQ ID Fungal Gene Library v20222 is compatible with Google Chrome v108.0.5359.73.
What are the operating system requirements for MicroSEQ ID Fungal Gene Library v2022?
The operating system requirements for MicroSEQ ID Fungal Gene Library v2022 are Windows 10, Enterprise LTSC 2018 Microsoft Corporation, 64 bit OS (Operating System), x64 Based Processor, Version-1809.
Which Windows OS (Operating System) is MicroSEQ ID Microbial Identification Software v4.0 compatible with?
MicroSEQ ID Microbial Identification Software v4.0 is compatible with 64-bit Windows 10 OS.
What are the computer requirements for MicroSEQ ID Microbial Identification Software v4.0?
The computer requirements for MicroSEQ ID Microbial Identification Software v4.0 are:
- Intel Core i7-8700 CPU at 3.20 Ghz, with 16 GB of RAM and a minimum of 465 GB hard disk space
- WIN 10 IoT 64-bit
Which MicroSeq ID (MSID) library versions is MicroSEQ ID Microbial Identification Software v4.0 compatible with?
MicroSEQ ID Microbial Identification Software v4.0 is compatible with MicroSEQ ID 16S rDNA Full Gene Library v2.1, MicroSEQ ID Fungal Gene Library v2022, MicroSEQ ID 16S rDNA 500 Library v2022, and MicroSEQ ID 16s rDNA 500 Supplemental Library v2022.
What is the product SKU for MicroSEQ ID Fungal Gene Library v2022?
The product SKU for MicroSEQ ID Fungal Gene Library v2022 is Cat. No. A57314.
For Research Use Only. Not for use in diagnostic procedures.