Learn More
Applied Biosystems™ MicroSEQ™ ID Fungal ITS Gene Library v1.0
Description
The MicroSEQ™ ID ITS Fungal Gene Library v1.0 is a curated collection of Internal Transcribed Spacer (ITS) sequences designed for accurate identification of fungal organisms. This library targets the highly polymorphic ITS region to enable high resolution species level identification of closely related or rare environmental fungi.
Summary:
- Target Region: Internal Transcribed Spacer (ITS)
- Library Size: 38,229 sequence entries
- Taxonomic Breadth: 3,686 genera and 22,265 fungal species
- Compatibility: Designed for use with MicroSEQ™ ID Analysis Software
Features
Comprehensive Coverage
Includes over 38,000 validated sequence entries spanning more than 22,000 fungal species, significantly reducing the likelihood of 'no-match' results for environmental and rare isolates.
High Discriminatory Power
The ITS region is highly polymorphic, enabling precise differentiation at the species level where conserved regions like the D2 (LSU) may lack sufficient resolution.
Curated and Validated Content
Sequence entries are meticulously curated to enable high standards of accuracy and consistency, offering reliable fungal identification for regulated environments.
Enhanced Identification Capability
Specifically designed to support the identification of closely related, rare, and environmental fungal isolates often encountered in industrial and environmental monitoring.
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Tube Centrifugation |
| Sequencing Type | DNA and RNA Sequencing |
| Product Line | MicroSEQ |
| Product Type | Library |
| Quantity | 1 Ea. |
| Software | MicroSEQ™ ID Analysis Software |
| Shipping Condition | Room Temperature |
| Target Organism Class | Fungi |
| Technique | SBT (Sequence-Based Typing) |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.