Learn More
Med Vet International Microserrated Bone Holding Forceps, X-Small, Serrated, 9cm, J0854A, Each
Catalog No. 5000584694
Encompass_Preferred
Extra Small Serrated Bone holding Forceps. 90mm (3 1/2") Manipulation of the long bones of cats and small dogs is difficult without crushing. The serrated jows of this forceps are contoured to maximize grip and support. The tips are sharp. They may be use
Catalog No. 50-005-84694 Supplier Med Vet International Supplier No. JOR854A
Extra Small Serrated Bone holding Forceps. 90mm (3 1/2") Manipulation of the long bones of cats and small dogs is difficult without crushing. The serrated jows of this forceps are contoured to maximize grip and support. The tips are sharp. They may be use