Learn More
Electron Microscopy Sciences MicroSlicer ZERO1N Fresh and Fixed Tissue Slicer
SDP
Catalog No. 501931223
Encompass_Preferred
MicroSlicers are the standard models for cutting live tissues. More convenient with stepping motor mounted on the elevating mechanism. Usability and performance enhanced by built-in retraction mechanism that lowers the specimen after each sectioning.
Catalog No. 50-193-1223 Supplier Electron Microscopy Sciences Supplier No. 93303
Add to Cart
Add to Cart
Request a Quote
MicroSlicers are the standard models for cutting live tissues. More convenient with stepping motor mounted on the elevating mechanism. Usability and performance enhanced by built-in retraction mechanism that lowers the specimen after each sectioning.