Learn More
Ibidi USA u-Slide 8 Well Glass Bottom, 90/cs
SDP
Catalog No. NC1754075
Encompass
A chambered coverslip with 8 wells and a #1.5H glass bottom, suitable for use in TIRF and single molecule applications. A cell culture chamber for observation of the sample through the coverslip-like bottom using high resolution microscopy.Surface Modification: #1.5H (170 µm +/- 5 µm) D 263 M Schott glass, sterilizedPcs./Box: 15 (individually packed)
This item is not returnable. View return policy
Catalog No. NC1754075 Supplier Ibidi USA Supplier No. 8082790
A chambered coverslip with 8 wells and a #1.5H glass bottom, suitable for use in TIRF and single molecule applications. A cell culture chamber for observation of the sample through the coverslip-like bottom using high resolution microscopy.Surface Modification: #1.5H (170 µm +/- 5 µm) D 263 M Schott glass, sterilizedPcs./Box: 15 (individually packed)