Learn More
Supertek™ Microslide™ - Body Defenses/Infection
GREENER_CHOICESCI_ED
Catalog No. S816729 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816729 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816729 Supplier Supertek™ Supplier No. T204
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. The Body Defenses/Infection Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. The Skin (200x)
    • 2. Epithelial Lining of Respiratory Tact (175x)/Isolated Cell from Wind Pipe (1400x)
    • 3. Pyloric Stomach (100x)/Eyelid (30x)
    • 4. White Blood Corpuscles (500x)/Phagocytosis (25,000x)
    • 5. Lymph Gland (40x)
    • 6. Tonsils and Adenoids (7x)
    • 7. The Liver (20x)
    • 8. Plasma Cell (37,000x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type The Virus Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile