Learn More
Supertek™ Microslide™ - Cell Nucleus
GREENER_CHOICESCI_ED
Catalog No. S816740 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816740 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816740 Supplier Supertek™ Supplier No. T248
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. The Cell Nucleus Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Animal Cell Nucleus - (900x)
    • 2. Plant Cell Nucleus - (1000x)
    • 3. Nuclear Reproduction - (100x)
    • 4. Human Blood Cells - (900x)
    • 5. Voluntary Muscle Cells - (900x)
    • 6. The Nuclear Membrane (T.E.M.) (100,000x)
    • 7. Nuclear Membrane Surface - (S.E.M.) - 100,000x)
    • 8. Endoplasmic Reticulum - (125,000x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Cell Culture Slide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile