Learn More
Supertek™ Microslide™ - Cells of Plants and Animals
GREENER_CHOICESCI_ED
Catalog No. S816724 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816724 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816724 Supplier Supertek™ Supplier No. T102
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate features. The Cells of Plants and Animals Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Cheek Lining Cells - Stained (900x)
    • 2. Onion Skin Cells - Stained (100x)
    • 3. Spirogyra Cells - (175x)
    • 4. Amoeba - (250x)
    • 5. Green Leaf Cells - Stained (275x)
    • 6. Wood Cells - Stained (40x)
    • 7. Bone Cells - Stained - (300x)
    • 8. Brain Cells - Stained - (300x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Cell Culture Slide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile