Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microslide™ Cleavage

Shop All Supertek Scientific Products


Click to view available options
Slide Type:
Photomicrograph Slide
Description
- Image titles on Microslide:
- 1. One-Cell Stage (320x)
- 2. Two-Cell Stage (320x)
- 3. Four-Cell Stage (320x)
- 4. Eight-Cell Stage (320x)
- 5. A Mass of Cells (320x)
- 6. The Blastula (320x)
- 7. Early Gastrula (160x)
- 8. Late Gastrula (240x)
- Comes with a Extensive Lesson Plan with Pocket to Store Microslide.
- Reproducible Worksheet and Storage Box with a Purchase of 15.
- Grades 9-12
Specifications
Specifications
| Classification | Animal Development |
| Material | Paper and Film |
| Type | Microslide |
| Grade | 9 to 12 |
| Includes | Microslide and Lesson Plan |
| Slide Type | Photomicrograph Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction