Learn More
Supertek™ Microslide™ - Digestive System
GREENER_CHOICESCI_ED
Catalog No. S816714 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816714 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
Catalog No. S816714 Supplier Supertek™ Supplier No. T067
Add to Cart
Add to Cart
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. The Digestive System Microslide™ is accompanied by a detailed lesson plan designed to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Salavary Gland
    • 2. Taste Buds (350x)
    • 3. Gullet (Esophagus) (4x)
    • 4. Smooth Muscle (700x)
    • 5. Juntion of Gullet and Stomach
    • 6. Stomach Lining and Gastric Glands
    • 7. Small Intestine
    • 8. Intestinal Glands and Villi (125x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Digestive System Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile