Learn More
Supertek™ Microslide™ - Fern Life Cycle
GREENER_CHOICESCI_ED
Catalog No. S816719 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816719 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816719 Supplier Supertek™ Supplier No. T084
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. This Fern Life Cycle Microslide™ is accompanied by a detailed lesson plan designed to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Sporophyte Leaf (43x)
    • 2. Spore Cases (80x)
    • 3. Germinating Spores (80x)
    • 4. Mature Gametophyte (20x)
    • 5. Sex Organs (240x)
    • 6. Archegone (480x)
    • 7. Embryo Sporophyte (12x)
    • 8. Young Sporophyte (3x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Fern Life Cycle Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile