Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Microslide™ Harmful and Helpful Bacteria
GREENER_CHOICESCI_ED

Catalog No. S00506 Shop All Supertek Scientific Products
Change view
Click to view available options
Slide Type:
Photomicrograph Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Type
S00506 Photomicrograph Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00506 Supplier Supertek™ Supplier No. T105
Add to Cart
Add to Cart

This Microslide™ Harmful and Helpful Bacteria consists of 8 images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. With a detailed lesson plan designed to stimulate, inform and question students about the topic.

  • Image titles on Microslide:
  • 1. 3-Types of Bacteria of Diphtheria - Stained (2,000x)
  • 2. Bacteria of Diphtheria - Stained - (2000x)
  • 3. Bacteria of Typhoid Fever - Stained (2,000x)
  • 4. Bacteria of Food Poisoning - Stained -(2,000)
  • 5. Bacteria of Potato Ring Rot - Stained (2000x)
  • 6. Bacteria of Blood Poisoning - Stained - (2,000x)
  • 7. Cheese Bacteria - Stained - (2,000)
  • 8. Vinegar Bacteria - Stained - (2,000x)
  • Comes with an Extensive Lesson Plan with a Pocket to store the Microscope.
  • Reproducible Worksheet and Storage Box with a purchase of 15.
  • Grades 6-8

Specifications

Classification Microbiology
Material Paper and Film
Type Microslide
Grade 6 to 8
Includes Microslide and Lesson Plan
Slide Type Photomicrograph Slide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.