Learn More
Supertek™ Microslide™ - Meiosis
GREENER_CHOICESCI_ED
Catalog No. S816720 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816720 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816720 Supplier Supertek™ Supplier No. T092
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. This Meiosis Microslide™ is accompanied by a detailed lesson plan designed to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Primary Oocyte-Tetrads Formed
    • 2. Primary Oocyte-Dyads Separated
    • 3. Primary Oocyte-Tetrads on Spindle
    • 4. Primary Oocyte-First Polar Body Forming (750x)
    • 5. Secondary Oocyte-Chromosomes Formed (750x)
    • 6. Mature Egg (750x)
    • 7. Pronucleus Stage(750x)
    • 8. Zygote Ready for First Cleavage (750x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Meiosis Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile