Learn More
Supertek™ Microslide™ - Micro Chromosomes and Genes in Action
GREENER_CHOICESCI_ED
Catalog No. S816718 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816718 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816718 Supplier Supertek™ Supplier No. T081
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope. Arrows and call outs help the student locate important features. This Micro Chromosomes and Genes in Action Microslide™ is accompanied by a detailed lesson plan designed to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. DNA Molecule Electron Micrograph - (55,000x)
    • 2. Coiling in Chromosomes w.m. - (2000x)
    • 3. Lamp Brush Chromosomes - Phase Contrast - (700x)
    • 4. Loop of Lamp Brush Chromosome Electron Micrographs - (9000x and 15,000x)
    • 5. Giant Chromosomes w.m. - (1000x)
    • 6. Chromosome Puffs Left - Stained (250x) Right - Tagged (250x)
    • 7.Nuclear Pores - Electron Micrograph - (50,000x)
    • 8 Ribosomes - Electron Micrograph - (85,000x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Cell Culture Slide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile