Learn More
Supertek™ Microslide™ - Monocots and Dicots Set
GREENER_CHOICESCI_ED
Catalog No. S816730 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816730 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816730 Supplier Supertek™ Supplier No. T210
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. The Monocots and Dicots Set Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. A Monocot Seed (20x)
    • 2. A Dicot Seed (40x)
    • 3. Petunia Seedling (10x)
    • 4. Monocot and Dicot Leaves
    • 5. Monocot or Dicot (50x)
    • 6. Monocot Dicot Stems (8x 34x)
    • 7. Dicot Stem (18x)
    • 8. Monocot Flower (6x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Monocots and Dicots Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile