Learn More
Supertek™ Microslide™ - Nerve and Muscle Action Set
GREENER_CHOICESCI_ED
Catalog No. S816735 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816735 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816735 Supplier Supertek™ Supplier No. T221
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate features. The Nerve and Muscle Action Set Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Single (100x)
    • 2. Synapse (left side - 100x) (right side - 3,000,000x)
    • 3. Axons (left side - x.s. - 400x) (right side - I.s. -200x)
    • 4. Myelinated Axons (600x)
    • 5. Nerve-Muscle Junction (400x)
    • 6. Sunmaptic Knobs - E.M. (300,000x)
    • 7. Striated Muscle Cell (1000x)
    • 8. Sarcomeres (left side relaxed - E.M. 45,000x) (right side - contracted - E.M. 31,000x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Nerve Muscle Action Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile