Learn More
Supertek™ Microslide™ - Respiratory System
GREENER_CHOICESCI_ED
Catalog No. S816717 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816717 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
Catalog No. S816717 Supplier Supertek™ Supplier No. T072
Add to Cart
Add to Cart
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. Respiratory System Microslide™ is accompanied by a detailed lesson plan designed to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Bronchial Tree (Human)
    • 2. Trachea and Gullet (6.5x)
    • 3. Cartlidges of the Chea (65x)
    • 4. Epithelial Lining of Respiratory Passage
    • 5. Air Sacs of the Lung (65x)
    • 6. The Capillary Networks (325x)
    • 7. Pigment Deposits in the Lung (325x)
    • 8. Mitochondrion (60,000x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Respiratory System Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile