Learn More
Supertek™ Microslide™ - Skeletal System
GREENER_CHOICESCI_ED
Catalog No. S816736 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816736 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816736 Supplier Supertek™ Supplier No. T223
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. The Skeletal System Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Bone - x.s. Stained (150x)
    • 2. Membranous - x.s. - Stained Bone (120x)
    • 3. Bone Marrow - Stained (Left Side Normal (400x)/Right side - Leukemia (500x)
    • 4. Bone Fracture-Arm-X-Ray (1/8x)
    • 5. Cartilage - x.s. - stained (69x)
    • 6. Arthritis-Hands(1/6x)/Feet-X-Ray (1/8x)
    • 7. Ligament at a Joint (1.5x)/fibers (200x)
    • 8. Muscle-Tendon-Bone Connetions (400x) (10x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Human Body Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile