Learn More
Supertek™ Microslide™ - Symbiosis Set
GREENER_CHOICESCI_ED
Catalog No. S816731 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816731 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816731 Supplier Supertek™ Supplier No. T212
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. The Symbiosis Set Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Trypanasomes in Rat Blook (1500x)
    • 2. Soybean Root Nodules - Stained - x.s. (100x)
    • 3. Nematode on Tomato Root (200x)
    • 4. Green Paramecium (200x)
    • 5. Rust on Willow Leaf (200x)
    • 6. Lichen (200x)
    • 7. Fungus on Fish (5x)
    • 8. Virus on Bacteria in Man (45,000x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile