Learn More
Description
- Image titles on Microslide:
- 1. Epithelial Cells (900x)
- 2. Cell Membrane (3500x)
- 3. Paramecium-(dying) (200x)
- 4. Cilia -(Locomotion) (1500x)
- 5. Blood Cells - (Structures) (26x)
- 6. Blood cells -(functions) (600x)
- 7. White Blood cell-(Recognition) (800x)
- 8. Onion Skin Cells (100x)
- Lesson Plan with Pocket to store Microslide
- Reproducible Worksheet and Storage Box with a purchase of 15
- Grades 1-8
Specifications
Specifications
| Classification | Cell Biology |
| Material | Paper and Film |
| Type | Microslide |
| Grade | 1 to 8 |
| Includes | Microslide and Lesson Plan |
| Slide Type | Photomicrograph Slide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

