Learn More
Supertek™ Microslide™ - The Five Kingdom
GREENER_CHOICESCI_ED
Catalog No. S816738 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816738 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816738 Supplier Supertek™ Supplier No. T241
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. The Five Kingdom Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Eugena - A classification puzzle (400x)
    • 2. The Kingdom Monera - Nitrogen Fixing Bacteria (2000x)
    • 3. The Kindgom Monera - Gleocapsa (1000x)
    • 4. The Kingdom Protista - Paramecium (100x)
    • 5. The Kingdom Protista - Spirocygra (150x)
    • 6. The Fungus Kingdom - The Mushroom (1/2x, 400x)
    • 7. The Plant Kingdom - Onion (xx0x) Elodea (150x)
    • 8. The Animal Kindom - (750x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile