Learn More
Supertek™ Microslide™ - The Kingdom Fungi Set
GREENER_CHOICESCI_ED
Catalog No. S816732 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816732 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816732 Supplier Supertek™ Supplier No. T214
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microslide™ strip consist of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. The Kingdom Fungi Set Microslide™ is accompanied by a detailed lesson plan to stimulate, inform, and question students.
  • Image titles on Microslide:
    • 1. Achyla ( Division Oomycota) (500x)
    • 2. Rhizopus (Division Zygomycota) (20x)
    • 3. Saccharomyces (Division Ascomycota )(500x)
    • 4. Peziza Division Ascomycota) x.s. (400x)
    • 5. Mushroom ( Division Basidiomycota) I.s.(1/2x)
    • 6. Mushroom (Division Basidiomycota) x.s.(400x)
    • 7. Epidermophyton Floccosum ( Division Deuteromycota) (500x)
    • 8. Lichen (Division Mycophycophyta) (25x)
  • Lesson Plan with Pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Microslide
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile