Learn More
Supertek™ Microslide™ - Ultra Structure of Animal Cell
GREENER_CHOICESCI_ED
Catalog No. S816700 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (LxW):
Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Catalog No. Dimensions (LxW)
S816700 Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
1 options
This item is not returnable. View return policy
Catalog No. S816700 Supplier Supertek™ Supplier No. T066
Add to Cart
Add to Cart
This item is not returnable. View return policy
One Microslide™ strip consists of images photographed through a microscope called photomicrographs. Arrows and call outs help the student locate important features. This Ultra Structure of Animal Cell Microslide™ is accompanied by a detailed lesson plan.
  • Image Titles on Microslide:
    • 1. Gland Cells (400x) - x.s.- (170x)
    • 2. Plasma Cell (37,000x)
    • 3. Nerve Cell Membrane (4,000,000x)
    • 4. Golgi Body and Vacuole (26,000x)
    • 5. Mitochondrion (124,000x)
    • 6. Centriole
    • 7. Lamp-Brush Chromosomes (700x)
    • 8. Muscle Cell Fibrils (45,000x)
  • Lesson Plan with pocket to store Microslide
  • Reproducible Worksheet with a purchase of 15
Specifications
Type Animal Tissue Microslide
Color Clear
Surface Treatment Untreated
Material Paper and Film
Dimensions (LxW) Lesson Plan 6.5 x 8 in., Slide 2 x 8 in.
Product Line Supertek Microslides
Sterility Non-sterile