Learn More
Abcam Microsome isolation kit, 50 Tests
Catalog No. 502205084
Encompass_Preferred
Microsome Isolation Kit (ab206995) provides a convenient and fast way to isolate microsomal fractions from animal tissues for downstream applications such as assessing CYP-mediated drug metabolism and xenobiotic biotransformation, and protein profiling of microsomal membrane proteins by SDS-PAGE and western blotting.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5084 Supplier Abcam Supplier No. AB20699550TEST
Microsome Isolation Kit (ab206995) provides a convenient and fast way to isolate microsomal fractions from animal tissues for downstream applications such as assessing CYP-mediated drug metabolism and xenobiotic biotransformation, and protein profiling of microsomal membrane proteins by SDS-PAGE and western blotting.
The product is subject to the following: Abcam Restricted Use Statement