Learn More
Sigma Aldrich Fine Chemicals Biosciences MicrospinTM S 300 HR Cytiv
Catalog No. 502904273
Encompass
MicroSpin(TM) columns are designed for the rapid purification of nucleic acids for use in a wide range of applications including desalting buffer exchange and removal of primers.
Catalog No. 50-290-4273 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE27513001
MicroSpin(TM) columns are designed for the rapid purification of nucleic acids for use in a wide range of applications including desalting buffer exchange and removal of primers.