Learn More
Sigma Aldrich Fine Chemicals Biosciences MicrospinTM S 400 HR Cytiv
Catalog No. 502904430
Encompass
MicroSpin(TM) columns are designed for the rapid purification of nucleic acids for use in a wide range of applications including desalting buffer exchange and removal of primers. Good product yield and purity is obtained with sample volumes from 25-100 mul and from nanogram to milligram quantities of DNA.
Catalog No. 50-290-4430 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE27514001
MicroSpin(TM) columns are designed for the rapid purification of nucleic acids for use in a wide range of applications including desalting buffer exchange and removal of primers. Good product yield and purity is obtained with sample volumes from 25-100 mul and from nanogram to milligram quantities of DNA.