Learn More
StatLab Microsporidia Control Slides
SDP
Catalog No. 22130476
Change view
Click to view available options
Quantity:
10 Slides/Box
Catalog No. Quantity
22-130-476 10 Slides/Box
1 options
This item is not returnable. View return policy
Catalog No. 22-130-476 Supplier StatLab Supplier No. 369
Add to Cart
Add to Cart
This item is not returnable. View return policy
Eliminate the need to make your own cultures for QC.
  • Microsporidia Control Slides contain high organism counts on positive wells and satisfy regulatory requirements for microsporidia
  • Positive well: Microsporidia in fecal flora
  • Negative well: fecal flora with yeast only
  • 10 slides/box
Specifications
Product Type Control Slide
Quantity 10 Slides/Box