Learn More
Braintree Scientific Microsurgical Instrument Kit,Instrument Set only
SDP
Catalog No. 501955232
Encompass
Microsurgical Instrument Kit - Includes DERF needle holder: grated jaws, 4000 teeth per square inch, 4.75" (12.1cm); Iris scissors: straight, 4" (10.2cm); SWISS jeweler style forceps: non-magnetic stainless steel, style 5, micro-fine, 4.375" (11.2cm); McPHERSON micro iris suturing forceps: straight, smooth platform 0.3mm x 5mm, no teeth, 3.5" (8.9cm); McPHERSON-VANNAS micro iris scissors: straight, sharp points, 3.25" (8.3cm); PlainWire eye speculum: blades 15mm wide, 2.5" (6.4cm); Probe: double ended, malleable, sterling, 4.5" (11.4cm); Storage cassette: 6.5" x 6.25" x 1.25"; Storage cassette mat **Sold for research purposes only and not for human use**
Catalog No. 50-195-5232 Supplier Braintree Scientific Supplier No. MSK6890NC
Microsurgical Instrument Kit - Includes DERF needle holder: grated jaws, 4000 teeth per square inch, 4.75" (12.1cm); Iris scissors: straight, 4" (10.2cm); SWISS jeweler style forceps: non-magnetic stainless steel, style 5, micro-fine, 4.375" (11.2cm); McPHERSON micro iris suturing forceps: straight, smooth platform 0.3mm x 5mm, no teeth, 3.5" (8.9cm); McPHERSON-VANNAS micro iris scissors: straight, sharp points, 3.25" (8.3cm); PlainWire eye speculum: blades 15mm wide, 2.5" (6.4cm); Probe: double ended, malleable, sterling, 4.5" (11.4cm); Storage cassette: 6.5" x 6.25" x 1.25"; Storage cassette mat **Sold for research purposes only and not for human use**