Learn More
Andwin Scientific MICROTAINR DIPOTASS EDTA 200/C
SDP
Catalog No. NC0399088
Encompass_Preferred
The BD Microtainer� Tubes are ideal for the collection, transport, and processing of skin puncture blood. This product features an integrated collection scoop, together with a wider diameter tube, which permit ease of collection and improved mixing. This product is sold in a case of 200 tubes with a stopper in the color of lavender.
This item is not returnable. View return policy
Catalog No. NC0399088 Supplier Andwin Scientific Supplier No. VT365974
The BD Microtainer� Tubes are ideal for the collection, transport, and processing of skin puncture blood. This product features an integrated collection scoop, together with a wider diameter tube, which permit ease of collection and improved mixing. This product is sold in a case of 200 tubes with a stopper in the color of lavender.