Learn More
Sirius Automation Inc MicroTasker Benchtop Lab Robot
SDP
Supplier:  Sirius Automation Inc MNT80101
Encompass
This Small-Footprint (requires only 2 feet of linear bench space) General Purpose Modular Laboratory Robot handles everyday tasks such as Analytical Weighing (4 or 5 place) Sample ID (1D or 2D barcodes) Sorting Dilutions Vortex Mixing Internal Standard Addition Standard Prep Un-Capping Re-Capping Transfers Aliquoting Liquid handling with sub-microliter accuracy Sample Capacity accommodates up to 8 microplate racks or custom equivalent) with your choice of disposables and labware Easy to use interface (21CFR11 compliance options) with LIMS compatible data management
Catalog No. 50-000-78212
Add to Cart
Request a Quote