Learn More
Braintree Scientific MicroTattooing Platform with Single Flexible LED Light
SDP
Catalog No. 501955414
Encompass
MicroTattooing Platform with Single Flexible LED Light can be used for surgery, micro manipulations, general micro-utility platform (i.e. organ inspections, dissections, etc.). Flex arm, non-obstructive rimless magnifying glass (main lens: 2.3 X; inset lens: 4.0 X). Non-skid feet for added stability. Universal input power supply (100-240 VAC, 50-60 Hz). Surgery Platform incorporates one flexible LED light which provides direct illumination, no shadows and no heat. Includes 1.5mL cup holders. Size: 6"x9" **Sold for research purposes only and not for human use**
Catalog No. 50-195-5414 Supplier Braintree Scientific Supplier No. NTS01120V
MicroTattooing Platform with Single Flexible LED Light can be used for surgery, micro manipulations, general micro-utility platform (i.e. organ inspections, dissections, etc.). Flex arm, non-obstructive rimless magnifying glass (main lens: 2.3 X; inset lens: 4.0 X). Non-skid feet for added stability. Universal input power supply (100-240 VAC, 50-60 Hz). Surgery Platform incorporates one flexible LED light which provides direct illumination, no shadows and no heat. Includes 1.5mL cup holders. Size: 6"x9" **Sold for research purposes only and not for human use**