Learn More
Andwin Scientific MICROTBE 1.5ML PRSTL 2500/CS
SDP
Catalog No. NC9940924
Encompass_Preferred
The 1.5ml pre-sterilized microtube does not open during boiling. It features ultrasmooth conical bottoms, frosted surface area, graduation marking, thin membrane in cap for easy syringe penetration and diamond polished molds.
This item is not returnable. View return policy
Catalog No. NC9940924 Supplier Andwin Scientific Supplier No. MCT150CS
The 1.5ml pre-sterilized microtube does not open during boiling. It features ultrasmooth conical bottoms, frosted surface area, graduation marking, thin membrane in cap for easy syringe penetration and diamond polished molds.