Learn More
Electron Microscopy Sciences Microtube 81-Well Storage Rack, Assorted Colors
SDP
Catalog No. 501899012
Encompass_Preferred
Durable polypropylene rack fits in standard 5.25" square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials. Features molded grid lines on the lid, imprinted coordinates on the bottom of base, and a molded reference point on the lid. Rack measures 5.125 x 5.125 x 1.75".
Color: Assorted Colors
Catalog No. 50-189-9012 Supplier Electron Microscopy Sciences Supplier No. 61600A
Durable polypropylene rack fits in standard 5.25" square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials. Features molded grid lines on the lid, imprinted coordinates on the bottom of base, and a molded reference point on the lid. Rack measures 5.125 x 5.125 x 1.75".
Color: Assorted Colors