Learn More
Electron Microscopy Sciences Microtube 81-Well Storage Rack, Blue
SDP
Catalog No. 501899013
Encompass_Preferred
Durable polypropylene rack fits in standard 5.25" square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials. Features molded grid lines on the lid, imprinted coordinates on the bottom of base, and a molded reference point on the lid. Rack measures 5.125 x 5.125 x 1.75".
Color: Blue
Catalog No. 50-189-9013 Supplier Electron Microscopy Sciences Supplier No. 61600B
Durable polypropylene rack fits in standard 5.25" square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials. Features molded grid lines on the lid, imprinted coordinates on the bottom of base, and a molded reference point on the lid. Rack measures 5.125 x 5.125 x 1.75".
Color: Blue