Learn More
Electron Microscopy Sciences BeadBug™ Microtube Homogenizer, 230V, EU Plug
SDP
Catalog No. 501931124
Encompass_Preferred
The BeadBug sets the performance standard for personal sized cell disruption and lysis instruments. Developed for samples too tough to be homogenized in a common lab vortexer, the BeadBug provides a convenient and economical alternative to the traditional, over-priced homogenizers.
With a foot print of just 7 x 8" and a capacity of up to three tubes, the BeadBug is the ideal homogenizing instrument for low to medium throughput laboratories.
Simultaneous homogenization takes place (often within 45 seconds) of up to 3 samples inside the disposable 2 ml screw cap microtubes. The optimized mixing motion causes rapid cell disruption through constant high velocity impact from the hardened micro-beads chosen specifically for your sample type.
Catalog No. 50-193-1124 Supplier Electron Microscopy Sciences Supplier No. 7262002
The BeadBug sets the performance standard for personal sized cell disruption and lysis instruments. Developed for samples too tough to be homogenized in a common lab vortexer, the BeadBug provides a convenient and economical alternative to the traditional, over-priced homogenizers.
With a foot print of just 7 x 8" and a capacity of up to three tubes, the BeadBug is the ideal homogenizing instrument for low to medium throughput laboratories.
Simultaneous homogenization takes place (often within 45 seconds) of up to 3 samples inside the disposable 2 ml screw cap microtubes. The optimized mixing motion causes rapid cell disruption through constant high velocity impact from the hardened micro-beads chosen specifically for your sample type.